Stampa…

9-(4-Iodophenyl)carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-(4-Iodophenyl)-9H-carbazole

I0961-1G I0961-5G
TCIAI0961-1GEA 138 EUR
TCIAI0961-1G TCIAI0961-5G
9-(4-Iodophenyl)carbazole ≥98.0% (by GC)
9-(4-Iodophenyl)carbazole
Formula: C₁₈H₁₂IN
Peso molecolare: 369,21 g/mol
MDL Number: MFCD11617970
CAS Number: 57103-15-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR