Stampa…

Imidocarb Dipropionate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3-Bis(3-(4,5-dihydro-1H-imidazol-2-yl)phenyl)urea dipropionate

I0957-1G I0957-5G
TCIAI0957-1GEA 50 EUR
TCIAI0957-1G TCIAI0957-5G
Imidocarb Dipropionate ≥98.0% (by HPLC, titration analysis)
Imidocarb Dipropionate
Formula: C₁₉H₂₀N₆O·₂C₃H₆O₂
Peso molecolare: 496,57 g/mol
MDL Number: MFCD00013161
CAS Number: 55750-06-6
EINECS: 259-791-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5228fc1b149d57df3b6ef0c704f7d9118b70ee8&vid=d5228fc1b149d57df3b6ef0c704f7d9118b70ee8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR