4-Isopropylthiophenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-(Isopropylthio)benzeneboronic acid, 4-(Isopropylthio)phenylboronic acid
I0950-1G
I0950-5G
TCIAI0950-1GEA
26
EUR
TCIAI0950-1G
TCIAI0950-5G
4-Isopropylthiophenylboronic acid (contains varying amounts of Anhydride)
4-Isopropylthiophenylboronic acid
Formula:
C₉H₁₃BO₂S Peso molecolare: 196,08 g/mol |
MDL Number:
MFCD03411947 CAS Number: 380427-38-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...