Stampa…

Isoquinoline-5-carbaldehyde ≥98.0% (by GC)

Fornitore: TCI
Attenzione

I0942-5G I0942-1G
TCIAI0942-5GEA 500 EUR
TCIAI0942-5G TCIAI0942-1G
Isoquinoline-5-carbaldehyde ≥98.0% (by GC)
Isoquinoline-5-carbaldehyde
Formula: C₁₀H₇NO
MDL Number: MFCD03412483
CAS Number: 80278-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc400f95d3cf96fcb9a911db609f5a43e8d0e87f&vid=fc400f95d3cf96fcb9a911db609f5a43e8d0e87f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR