Isobutylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
I0935-1G
I0935-5G
TCIAI0935-1GEA
54
EUR
TCIAI0935-1G
TCIAI0935-5G
Isobutylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Isobutylamine hydroiodide
Formula:
C₄H₁₁N Peso molecolare: 201,05 g/mol |
CAS Number:
205508-75-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...