Stampa…

6-Iodo-1H-indazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Iodoindazole

I0933-1G I0933-5G
TCIAI0933-1GEA 69 EUR
TCIAI0933-1G TCIAI0933-5G
6-Iodo-1H-indazole ≥98.0% (by GC)
6-Iodo-1H-indazole
Formula: C₇H₅IN₂
Peso molecolare: 244,0300 g/mol
MDL Number: MFCD04114695
CAS Number: 261953-36-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac6adbd0bd520a9434fd0eaadd25c28e2b5a2d14&vid=ac6adbd0bd520a9434fd0eaadd25c28e2b5a2d14&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 207.0 to 211.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR