(1-Indolyl)acetic acid ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Indole-1-acetic Acid, 2-(1H-Indol-1-yl)acetic acid
I0932-1G
I0932-5G
TCIAI0932-1GEA
29
EUR
TCIAI0932-1G
TCIAI0932-5G
(1-Indolyl)acetic acid ≥97.0% (by titrimetric analysis)
(1-Indolyl)acetic acid
Formula:
C₁₀H₉NO₂ Peso molecolare: 175,19 g/mol |
MDL Number:
MFCD00047262 CAS Number: 24297-59-4 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 168.0 to 172.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...