Isopropyl methanesulphonate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
I0914-1G
I0914-5G
TCIAI0914-1GEA
42
EUR
TCIAI0914-1G
TCIAI0914-5G
Isopropyl methanesulphonate ≥98.0% (by GC)
Isopropyl methanesulphonate
Formula:
C₄H₁₀O₃S Peso molecolare: 138,181 g/mol Punto di ebollizione: 67°C Punto di fusione: 7°C Flash Pt: 104°C |
MDL Number:
MFCD00047802 CAS Number: 926-06-7 EINECS: 213-132-0 UN: 2810 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...