Stampa…

2-Metilpropene ∼15% in THF

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Methylpropene, Isobutene, Isobutylene

I0911-100ML I0911-500ML
TCIAI0911-100MLEA 25 EUR
TCIAI0911-100ML TCIAI0911-500ML
2-Metilpropene ∼15% in THF
2-Metilpropene
Formula: C₄H₈
Peso molecolare: 56,11 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00008898
CAS Number: 115-11-7
EINECS: 204-066-3
UN: 1993
ADR: 3,II
Merck Index: 14,05141
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae0ab1b2f0e0764a78f04c29bdd934e23307eecb&vid=ae0ab1b2f0e0764a78f04c29bdd934e23307eecb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 14.0 to 16.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR