Stampa…

Imatinib ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide

I0906-100MG I0906-1G
TCIAI0906-100MGEA 45 EUR
TCIAI0906-100MG TCIAI0906-1G
Imatinib ≥98.0% (by HPLC)
Imatinib
Formula: C₂₉H₃₁N₇O
Peso molecolare: 493,62 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD05662257
CAS Number: 152459-95-5
Merck Index: 14,04902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75c75ca8d5a14a19b6719a13ce9284150a3523b7&vid=75c75ca8d5a14a19b6719a13ce9284150a3523b7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 207.0 to 211.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR