Stampa…

Iproniazid ≥96.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Isonicotinic acid 2-Isopropylhydrazide, 1-Isonicotinoyl-2-isopropylhydrazine

I0893-200MG I0893-1G
TCIAI0893-200MGEA 30 EUR
TCIAI0893-200MG TCIAI0893-1G
Iproniazid ≥96.0% (by HPLC, titration analysis)
Iproniazid
Formula: C₉H₁₃N₃O
Peso molecolare: 179,22 g/mol
MDL Number: MFCD00153819
CAS Number: 54-92-2
EINECS: 200-218-8
Merck Index: 14,05078
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a38bc076c5b6cc8f2768dd3afb7d580125ec413&vid=1a38bc076c5b6cc8f2768dd3afb7d580125ec413&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 109.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR