(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(S)-2-Hydroxypropionic acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, (S)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2-hydroxypropanoate, L-Menthyl L-Lactate
I0889-5G
I0889-25G
TCIAI0889-5GEA
27
EUR
TCIAI0889-5G
TCIAI0889-25G
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate ≥98.0% (by GC)
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate
Formula:
C₁₃H₂₄O₃ Peso molecolare: 228,33 g/mol |
MDL Number:
MFCD27977194 CAS Number: 61597-98-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Specific rotation [a]20/D | -77.0 to -83.0 deg (C=5, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...