Stampa…

3-Isopropylphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Isopropylbenzeneboronic acid

I0887-1G I0887-5G
TCIAI0887-1GEA 113 EUR
TCIAI0887-1G TCIAI0887-5G
3-Isopropylphenylboronic acid (contains varying amounts of Anhydride)
3-Isopropylphenylboronic acid
Formula: C₉H₁₃BO₂
Peso molecolare: 164,01 g/mol
MDL Number: MFCD01074634
CAS Number: 216019-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79bf98d60320b9b8677b705215a69416b5ece61e&vid=79bf98d60320b9b8677b705215a69416b5ece61e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR