Stampa…

3-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Isophthalaldehydic acid, 3-Carboxybenzaldehyde

I0851-1G I0851-5G
TCIAI0851-1GEA 37 EUR
TCIAI0851-1G TCIAI0851-5G
3-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Formylbenzoic acid
Formula: C₈H₆O₃
Peso molecolare: 150.13 g/mol
Punto di ebollizione: 306
Punto di fusione: 175
MDL Number: MFCD00039575
CAS Number: 619-21-6
EINECS: 210-585-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd32d906b96ba91041b2e35df7572cc53eca52fb&vid=fd32d906b96ba91041b2e35df7572cc53eca52fb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR