Stampa…

Ivabradine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI

I0847-200MG I0847-1G
TCIAI0847-200MGEA 92 EUR
TCIAI0847-200MG TCIAI0847-1G
Ivabradine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ivabradine hydrochloride
Formula: C₂₇H₃₆N₂O₅
Punto di fusione: 140°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00929899
CAS Number: 148849-67-6
Merck Index: 14,05247
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28338227f8c90a8576df0a48d4477ca6bcd37334&vid=28338227f8c90a8576df0a48d4477ca6bcd37334&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 97.0 %
Specific rotation [a]20/D +6.0 to +8.5 deg (C=1, DMSO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR