Stampa…

Isoproturon ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4-Isopropylphenyl)-1,1-dimethylurea

I0843-5G I0843-25G I0843-1G
TCIAI0843-5GEA 96 EUR
TCIAI0843-5G TCIAI0843-25G TCIAI0843-1G
Isoproturon ≥98.0% (by HPLC, total nitrogen)
Isoproturon
Formula: C₁₂H₁₈N₂O
Peso molecolare: 206,29 g/mol
MDL Number: MFCD00078684
CAS Number: 34123-59-6
EINECS: 251-835-4
UN: 3077
ADR: 9,III
Merck Index: 14,05218
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3405a066103a0706a796992f142ba3b7282b8f1&vid=21f590e3175fb364ef5043c4afef79f06cfb0345&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR