4-Iodobenzylamine ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
(4-Iodophenyl)methanamine
Formula:
C₇H₈IN Peso molecolare: 233.05 g/mol Punto di fusione: 45 |
MDL Number:
MFCD00047933 CAS Number: 39959-59-6 UN: 3259 ADR: 8 III |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...