Stampa…

Isoliquiritigenin ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one, 2',4,4'-Trihydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one

I0822-100MG I0822-1G
TCIAI0822-100MGEA 17 EUR
TCIAI0822-100MG TCIAI0822-1G
Isoliquiritigenin ≥97.0% (by HPLC, titration analysis)
Isoliquiritigenin
Formula: C₁₅H₁₂O₄
Peso molecolare: 256,26 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00075907
CAS Number: 961-29-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 197.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR