4-Iodo-1-methylpyrazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
4-Iodo-1-methyl-1H-pyrazole-5-carboxylic acid
Formula:
C₅H₅IN₂O₂ Punto di fusione: 242°C |
MDL Number:
MFCD00461121 CAS Number: 75092-30-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...