Isopropyl 2-methylbutyrate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methylbutyric Acid Isopropyl Ester
I0736-25ML
I0736-500ML
TCIAI0736-25MLEA
0
EUR
TCIAI0736-25ML
TCIAI0736-500ML
Isopropyl 2-methylbutyrate ≥98.0% (by GC)
Isopropyl 2-methylbutyrate
Formula:
C₈H₁₆O₂ Peso molecolare: 144,21 g/mol Punto di ebollizione: 144°C Flash Pt: 32°C |
MDL Number:
MFCD00085203 CAS Number: 66576-71-4 EINECS: 266-411-4 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...