Stampa…

Itraconazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Oriconazole

I0732-25G I0732-5G
TCIAI0732-25GEA 290 EUR
TCIAI0732-25G TCIAI0732-5G
Itraconazole ≥98.0% (by HPLC, titration analysis)
Itraconazole
Formula: C₃₅H₃₈Cl₂N₈O₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00870168
CAS Number: 84625-61-6
Merck Index: 14,05245

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR