Stampa…

Irsogladine maleate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: 6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine maleate, 2,4-Diamino-6-(2,5-dichlorophenyl)-1,3,5-triazine Maleate

I0719-1G I0719-5G
TCIAI0719-1GEA 79 EUR
TCIAI0719-1G TCIAI0719-5G
Irsogladine maleate ≥98.0% (by HPLC, titration analysis)
Irsogladine maleate
Formula: C₉H₇Cl₂N₅
MDL Number: MFCD00873566
CAS Number: 84504-69-8
Merck Index: 14,05102

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR