N2-Isobutyryl-2'-deoxyguanosine ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
N-(9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide
I0700-1G
I0700-100MG
TCIAI0700-1GEA
226
EUR
TCIAI0700-1G
TCIAI0700-100MG
N2-Isobutyryl-2'-deoxyguanosine ≥98.0% (by HPLC, total nitrogen)
N2-Isobutyryl-2'-deoxyguanosine
Formula:
C₁₄H₁₉N₅O₅ Peso molecolare: 337,34 g/mol Punto di fusione: 300°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00010060 CAS Number: 68892-42-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...