Stampa…

Isobornyl methacrylate ≥85.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Attenzione

Synonyms: rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate, (1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate(relative), Methacrylic acid Isobornyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

I0617-25G I0617-500G
TCIAI0617-25GEA 17 EUR
TCIAI0617-25G TCIAI0617-500G
Isobornyl methacrylate ≥85.0% (by GC) stabilizzato
Isobornyl methacrylate
Formula: C₁₄H₂₂O₂
Peso molecolare: 222,33 g/mol
Punto di ebollizione: 129°C
Flash Pt: 144°C
MDL Number: MFCD00081070
CAS Number: 7534-94-3
EINECS: 231-403-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR