Stampa…

Tris[3-(trimethoxysilyl)propyl] Isocyanurate ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3,5-Tris(3-(trimethoxysilyl)propyl)-1,3,5-triazinane-2,4,6-trione, Isocyanuric acid tris[3-(trimethoxysilyl)propyl] ester

I0602-5G I0602-25G
TCIAI0602-5GEA 18 EUR
TCIAI0602-5G TCIAI0602-25G
Tris[3-(trimethoxysilyl)propyl] Isocyanurate ≥95.0% (by GC)
Tris[3-(trimethoxysilyl)propyl] Isocyanurate
Formula: C₂₁H₄₅N₃O₁₂Si₃
Peso molecolare: 615,86 g/mol
Punto di ebollizione: 236°C
Flash Pt: 102°C
MDL Number: MFCD00054746
CAS Number: 26115-70-8
EINECS: 247-465-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f78e4d77eee811ea71dea8877333c486deead68e&vid=f78e4d77eee811ea71dea8877333c486deead68e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR