(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(R)-(-)-3-Propionyl-4-isopropyl-2-oxazolidinone, (R)-4-Isopropyl-3-propionyloxazolidin-2-one
I0594-1G
I0594-5G
TCIAI0594-1GEA
83
EUR
TCIAI0594-1G
TCIAI0594-5G
(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥98.0% (by GC)
(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone
Formula:
C₉H₁₅NO₃ Peso molecolare: 185,22 g/mol |
MDL Number:
MFCD00269691 CAS Number: 89028-40-0 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Specific rotation [a]20/D | -87.0 to -92.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...