Stampa…

(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (S)-4-Isopropyl-3-propionyloxazolidin-2-one

I0573-1G I0573-5G
TCIAI0573-1GEA 74 EUR
TCIAI0573-1G TCIAI0573-5G
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥97.0% (by GC)
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone
Formula: C₉H₁₅NO₃
Peso molecolare: 185,22 g/mol
MDL Number: MFCD00043401
CAS Number: 77877-19-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Optical purity (LC) min. 98.0 ee%
Specific rotation [a]20/D +87.0 to +91.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR