4-Iodo-4'-nitrobiphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Iodo-4'-nitro-1,1'-biphenyl
I0537-1G
I0537-5G
TCIAI0537-1GEA
26
EUR
TCIAI0537-1G
TCIAI0537-5G
4-Iodo-4'-nitrobiphenyl ≥98.0% (by GC)
4-Iodo-4'-nitrobiphenyl
Formula:
C₁₂H₈INO₂ Peso molecolare: 325,11 g/mol |
MDL Number:
MFCD00024698 CAS Number: 29170-08-9 EINECS: 249-480-5 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 214.0 to 218.0 deg-C |
Solubility in N,N-DMF | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...