Stampa…

L-Isoleucine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: H-Ile-OMe·HCl, H-Ile-OMe.HCl

I0522-1G I0522-5G
TCIAI0522-1GEA 16 EUR
TCIAI0522-1G TCIAI0522-5G
L-Isoleucine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-Isoleucine methyl ester hydrochloride
Formula: C₇H₁₅NO₂
Peso molecolare: 181,66 g/mol
MDL Number: MFCD00038911
CAS Number: 18598-74-8
EINECS: 242-437-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 97.0 to 100.0 deg-C
Specific rotation [a]20/D +25.0 to +28.0 deg (C=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR