Methyl (2S,3S)-2-isocyanato-3-methylvalerate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(2S,3S)-2-Isocyanato-3-methylvaleric Acid Methyl Ester
I0471-5G
I0471-1G
TCIAI0471-5GEA
186
EUR
TCIAI0471-5G
TCIAI0471-1G
Methyl (2S,3S)-2-isocyanato-3-methylvalerate ≥98.0% (by GC)
Methyl (2S,3S)-2-isocyanato-3-methylvalerate
Formula:
C₈H₁₃NO₃ Peso molecolare: 171,20 g/mol |
MDL Number:
MFCD00191534 CAS Number: 120219-17-2 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -4.0 to -6.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...