Stampa…

Diethyl-(S)-(-)-2-isocyanatoglutarate ≥97.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: (S)-(-)-2-Isocyanatoglutaric Acid Diethyl Ester, (S)-(-)-2-Isocyanatopentanedioic Acid Diethyl Ester, Diethyl (S)-(-)-2-Isocyanatopentanedioate, Diethyl (S)-2-isocyanatopentanedioate

I0468-1G I0468-5G
TCIAI0468-1GEA 94 EUR
TCIAI0468-1G TCIAI0468-5G
Diethyl-(S)-(-)-2-isocyanatoglutarate ≥97.0% (by GC)
Diethyl-(S)-(-)-2-isocyanatoglutarate
Formula: C₁₀H₁₅NO₅
Punto di ebollizione: 151°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00191663
CAS Number: 145080-95-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Specific rotation [a]20/D -41.0 to -46.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR