Stampa…

Tetrabutylammonium dihydrogen phosphate 0.5 mol/L in water per cromatografia

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

I0367-10ML I0367-100ML
TCIAI0367-10MLEA 19 EUR
TCIAI0367-10ML TCIAI0367-100ML
Tetrabutylammonium dihydrogen phosphate 0.5 mol/L in water per cromatografia
Tetrabutylammonium dihydrogen phosphate
Formula: C₁₆H₃₈NO₄P
Peso molecolare: 339,46 g/mol
MDL Number: MFCD00064526
CAS Number: 5574-97-0
EINECS: 226-947-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f6d560c8aa2c3d4f50e191c939ef6914d5956c6&vid=59ef918ee2a231ffcbb75867090cc3b57f6180d3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless clear liquid
Absorbance (230nm) max. 0.050
Absorbance (254nm) max. 0.010
pH 7.1 to 7.7 (25 deg-C)
Bromide max. 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR