Stampa…

Butyl Isocyanatoacetate ≥97.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: Isocyanatoacetic Acid Butyl Ester

I0358-25G I0358-5G
TCIAI0358-25GEA 325 EUR
TCIAI0358-25G TCIAI0358-5G
Butyl Isocyanatoacetate ≥97.0% (by GC)
Butyl Isocyanatoacetate
Formula: C₇H₁₁NO₃
Punto di ebollizione: 79°C
Flash Pt: 88°C
MDL Number: MFCD00041743
CAS Number: 17046-22-9
EINECS: 241-114-2
UN: 2206
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR