Diethyl iminodiacetate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
I0326-25ML
I0326-5ML
TCIAI0326-25MLEA
78
EUR
TCIAI0326-25ML
TCIAI0326-5ML
Diethyl iminodiacetate ≥98.0% (by GC)
Diethyl iminodiacetate
Formula:
C₈H₁₅NO₄ Punto di ebollizione: 122°C |
MDL Number:
MFCD00041925 CAS Number: 6290-05-7 EINECS: 228-533-6 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...