DL-Isoserine ≥95.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₃H₇NO₃ |
MDL Number:
MFCD00008138 CAS Number: 565-71-9 EINECS: 918-962-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...