Stampa…

Triallyl isocyanurate ≥96.0% (by GC) stabilizzato

Fornitore: TCI
Pericolo

Synonyms: 1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, Isocyanuric Acid Triallyl Ester, 1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione

Stabilization: stabilizzato con BHT/Ionolo

I0279-25G I0279-500G I0279-100G
TCIAI0279-25GEA 16 EUR
TCIAI0279-25G TCIAI0279-500G TCIAI0279-100G
Triallyl isocyanurate ≥96.0% (by GC) stabilizzato
Triallyl isocyanurate
Formula: C₁₂H₁₅N₃O₃
Punto di ebollizione: 140°C
MDL Number: MFCD00006554
CAS Number: 1025-15-6
EINECS: 213-834-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a69644817b29edad0968811e552e96942ffe6fc&vid=9a69644817b29edad0968811e552e96942ffe6fc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR