Monobutyl itaconate ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Butyl Methylenesuccinate, beta-Butyl Itaconate, Itaconic Acid Monobutyl Ester
I0267-25G
I0267-500G
TCIAI0267-25GEA
40
EUR
TCIAI0267-25G
TCIAI0267-500G
Monobutyl itaconate ≥95.0% (by GC)
Monobutyl itaconate
Formula:
C₉H₁₄O₄ Peso molecolare: 186,21 g/mol |
MDL Number:
MFCD00046533 CAS Number: 6439-57-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...