Stampa…

2-(Isopropylamino)ethanol ≥99.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-Isopropylethanolamine

I0227-25ML I0227-500ML I0227-100ML
TCIAI0227-25MLEA 25 EUR
TCIAI0227-25ML TCIAI0227-500ML TCIAI0227-100ML
2-(Isopropylamino)ethanol ≥99.0% (by GC, titration analysis)
2-(Isopropylamino)ethanol
Formula: C₅H₁₃NO
Peso molecolare: 103,17 g/mol
Punto di ebollizione: 171°C
Flash Pt: 76°C
MDL Number: MFCD00041755
CAS Number: 109-56-8
EINECS: 203-681-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR