Indigotrisulphonic acid potassium salt >55.0% (by HPLC)
Fornitore: TCI
Formula:
C₁₆H₇K₃N₂O₁₁S₃ |
MDL Number:
MFCD00013160 CAS Number: 67627-18-3 |
Risultati del test di specificazione
Appearance | Gray to Dark blue to Black powder to crystal |
Purity (HPLC) | min. 55,0 area% |
Suitability for Redox reagent | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...