2-Ethylhexyl vinyl ether ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Isooctyl Vinyl Ether
I0148-10G
I0148-25G
TCIAI0148-10GEA
94
EUR
TCIAI0148-10G
TCIAI0148-25G
2-Ethylhexyl vinyl ether ≥97.0% (by GC)
2-Ethylhexyl vinyl ether
Formula:
C₁₀H₂₀O Peso molecolare: 156,27 g/mol Punto di ebollizione: 177°C Flash Pt: 57°C |
MDL Number:
MFCD00053800 CAS Number: 103-44-6 EINECS: 203-111-4 UN: 3271 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...