Stampa…

Isopentylamine hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Methylbutan-1-amine hydrochloride, Isopentylammonium chloride

I0083-1G I0083-5G
TCIAI0083-1GEA 32 EUR
TCIAI0083-1G TCIAI0083-5G
Isopentylamine hydrochloride ≥98.0% (by titrimetric analysis)
Isopentylamine hydrochloride
Formula: C₅H₁₃N·HCl
Peso molecolare: 123,62 g/mol
MDL Number: MFCD00050684
CAS Number: 541-23-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR