Stampa…

(2-Hydroxy-5-methylphenyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: MeTAPS-I

H1749-1G H1749-5G
TCIAH1749-1GEA 50 EUR
TCIAH1749-1G TCIAH1749-5G
(2-Hydroxy-5-methylphenyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
(2-Hydroxy-5-methylphenyl)triphenylphosphonium iodide
Formula: C₂₅H₂₂IOP
Peso molecolare: 496.33 g/mol
CAS Number: 2005487-70-5

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Appearance White to Almost white powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR