(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
MeTAPS-Br
H1748-1G
H1748-5G
TCIAH1748-1GEA
50
EUR
TCIAH1748-1G
TCIAH1748-5G
(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(2-Hydroxy-5-methylphenyl)triphenylphosphonium bromide
Formula:
C₂₅H₂₂BrOP Peso molecolare: 449.33 g/mol |
CAS Number:
2005487-65-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...