Stampa…

4-(Hexyloxy)-2,3,6-trimethylphenol ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: HTHQ, HX-1171, 1-O-Hexyl-2,3,5-trimethylhydroquinone

H1741-250MG H1741-50MG
TCIAH1741-250MGEA 356 EUR
TCIAH1741-250MG TCIAH1741-50MG
4-(Hexyloxy)-2,3,6-trimethylphenol ≥95.0% (by GC)
4-(Hexyloxy)-2,3,6-trimethylphenol
Formula: C₁₅H₂₄O₂
Peso molecolare: 236,36 g/mol
MDL Number: MFCD00797612
CAS Number: 148081-72-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 95.0 %
Melting point 73.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR