2-(2-Hydroxyphenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2-(1H-Benzo[d]imidazol-2-yl)phenol, 2-(2′-Hydroxyphenyl)benzimidazole
H1736-25G
H1736-5G
TCIAH1736-25GEA
272
EUR
TCIAH1736-25G
TCIAH1736-5G
2-(2-Hydroxyphenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyphenyl)-1H-benzimidazole
Formula:
C₁₃H₁₀N₂O Peso molecolare: 210,24 g/mol |
MDL Number:
MFCD00022668 CAS Number: 2963-66-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 242.0 to 246.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...