(S)-1-(Pyridin-4-yl)ethanol ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-alpha-Methyl-4-pyridinemethanol, (S)-1-(4-Pyridyl)ethanol, (S)-4-(1-Hydroxyethyl)pyridine
H1734-1G
H1734-5G
TCIAH1734-1GEA
95
EUR
TCIAH1734-1G
TCIAH1734-5G
(S)-1-(Pyridin-4-yl)ethanol ≥97.0% (by GC, titration analysis)
(S)-1-(Pyridin-4-yl)ethanol
Formula:
C₇H₉NO Peso molecolare: 123,155 g/mol |
MDL Number:
MFCD00077866 CAS Number: 54656-96-1 |
Risultati del test di specificazione
Appearance | White to Light red to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Purity (GC) | min. 97.0 % |
Optical purity (GC) | min. 98.0 ee% |
Melting point | 67.0 to 71.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...