2-Heptyl-1,3-dioxolane ≥97.0% (by GC)
Fornitore: TCI
Formula:
C₁₀H₂₀O₂ Punto di ebollizione: 97°C |
MDL Number:
MFCD00216992 CAS Number: 4359-57-3 EINECS: 224-438-9 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...