2-Hydroxybutyl methacrylate (mixture of isomers) ≥97.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
Synonyms:
Methacrylic acid 2-Hydroxybutyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
H1714-25G
H1714-500G
TCIAH1714-25GEA
16
EUR
TCIAH1714-25G
TCIAH1714-500G
2-Hydroxybutyl methacrylate (mixture of isomers) ≥97.0% (by GC) stabilizzato
2-Hydroxybutyl methacrylate (mixture of isomers)
Formula:
C₈H₁₄O₃ Peso molecolare: 158,20 g/mol Punto di ebollizione: 98°C |
MDL Number:
MFCD19707078 CAS Number: 13159-51-8 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % (total of isomer) |
Acid Value | max. 2.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...