Hesperadin ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
(Z)-N-[2-Oxo-3-[phenyl[[4-(piperidin-1-ylmethyl)phenyl]amino]methylene]indolin-5-yl]ethanesulphonamide, (Z)-N-(2-Oxo-3-(phenyl((4-(piperidin-1-ylmethyl)phenyl)amino)methylene)indolin-5-yl)ethanesulphonamide
This product is unavailable in the U.S.
Formula:
C₂₉H₃₂N₄O₃S Peso molecolare: 516,65 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD18074526 CAS Number: 422513-13-1 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 225.0 to 230.0 deg-C |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...