N,N'-(Hexane-1,6-diyl)bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide] ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
Formula:
C₄₀H₆₄N₂O₄ |
MDL Number:
MFCD00134697 CAS Number: 23128-74-7 EINECS: 245-442-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...